Objectives for Rebel Microscopes
Plan achromat, N.A. 0,65, 0,4 mm
Catalog # ECHLRMAG0402
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Specifications
- Magnification:40×
- Description:Plan achromat, N.A. 0,65
- Seq:09
- Working distance:0,4 mm