Objectives for Rebel Microscopes
Plan achromat, N.A. 0,4, 1,6 mm
Catalog # ECHLRMAG0202
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
Specifications
- Magnification:20×
- Description:Plan achromat, N.A. 0,4
- Seq:16
- Working distance:1,6 mm
- Cat. No.:ECHLRMAG0202