Order Entry
Germany
Orders LinkContactUsLinkComponent
1,4-Dioxane solution, NMR reference standard, Supelco®

1,4-Dioxane solution, NMR reference standard, Supelco®

1,4-Dioxane solution, 40% in benzene-d6 (99.6 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., 1
Catalog # 736171-8IN
Supplier:   Merck
1,4-Dioxane solution, NMR reference standard, Supelco®
Catalog # 736171-8IN
Supplier:  Merck
CAS Number:  
1,4-Dioxane solution, NMR reference standard, Supelco®
1,4-Dioxane solution, NMR reference standard, Supelco®

1,4-Dioxane solution, 40% in benzene-d6 (99.6 atom % D), NMR reference standard, NMR tube size 5 mm x 8 in., 1

CAS Number:  
Price
355,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications

Specifications

  • Concentration:
    40% in benzene-d6 (99.6 atom % D)
  • Pk:
    1
  • Standards:
    NMR reference standard
  • Description:
    1,4-Dioxane solution
  • Pk Info:
    NMR tube size 5 mm x 8 in.
  • Cat. No.:
    736171-8IN
  • Description:
    1,4-Dioxane solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.