ContactUsLinkComponent
Multiple Distributors for Barrels EX, Screw cap for barrels with female thread and body with GL-threaded necks made of conductive PTFE-EX. Bore with female thread M5x6 for connecting a grounding cable. GL-threaded necks for inserting a tubing with a 1 * 1 items
Catalog # BOHLD693-14
Supplier:  Bohlender
undefined
Multiple Distributors for Barrels EX, Screw cap for barrels with female thread and body with GL-threaded necks made of conductive PTFE-EX. Bore with female thread M5x6 for connecting a grounding cable. GL-threaded necks for inserting a tubing with a 1 * 1 items
Catalog # BOHLD693-14
Supplier:  Bohlender