ContactUsLinkComponent
Rotor P-MRK, Suitable for BOLA Magnetic Stirrer Heads P-MRK with Ground Joint ns29/32 for shaft O. D. 8 mm, Cat. No. C 520-24 as wel l as BOLA Magnetic Stirrer Heads P-MRK with Ground Joint ns29/32 or ns45/40 for shaft O. D. 10 mm, Cat. No. C 520-28 1 * 1 items
Catalog # BOHLC935-01
Supplier:  Bohlender
undefined
Rotor P-MRK, Suitable for BOLA Magnetic Stirrer Heads P-MRK with Ground Joint ns29/32 for shaft O. D. 8 mm, Cat. No. C 520-24 as wel l as BOLA Magnetic Stirrer Heads P-MRK with Ground Joint ns29/32 or ns45/40 for shaft O. D. 10 mm, Cat. No. C 520-28 1 * 1 items
Catalog # BOHLC935-01
Supplier:  Bohlender