ContactUsLinkComponent
KL (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human Klotho in cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
  ABNOKA5480
 :  Abnova
KL (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human Klotho in cell culture supe rnates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
  ABNOKA5480
 :  Abnova
Frequently Bought Together