ContactUsLinkComponent
HWS flat flange cover, DN 150s, centre socket NS 29/32 side socket 2 x NS 29/32 straight, 2 x NS 29/32 angled +++flange acc. DIN 12.214+++ Customs No. 70172000 1 * 1 items
Catalog # HWSA3315051S
Supplier:  HWS LABORTECHNIK
HWS flat flange cover, DN 150s, centre socket NS 29/32 side socket 2 x NS 29/32 straight, 2 x NS 29/32 angled +++flange acc. DIN 12.214+++ Customs No. 70172000 1 * 1 items
Catalog # HWSA3315051S
Supplier:  HWS LABORTECHNIK