ContactUsLinkComponent
TemperatureProbes PT 1000 Lemo, One measuring sensor PT 1000 in a PTFE-encapsulated stainless steel tube (1.4571). Temperatureprobe Ø 8mm, tip Ø 6mm, collar ring Ø 12mm. With white PFA-coated cable (length: 1,5m) and mounted Lemo® plug size 1, 2-wi 1 * 1 items
Catalog # BOHLP1962-30
Supplier:  Bohlender
undefined
TemperatureProbes PT 1000 Lemo, One measuring sensor PT 1000 in a PTFE-encapsulated stainless steel tube (1.4571). Temperatureprobe Ø 8mm, tip Ø 6mm, collar ring Ø 12mm. With white PFA-coated cable (length: 1,5m) and mounted Lemo® plug size 1, 2-wi 1 * 1 items
Catalog # BOHLP1962-30
Supplier:  Bohlender