Order Entry
Germany
Orders LinkContactUsLinkComponent
Rat recombinant CNTF

Rat recombinant CNTF

500 µG
Catalog # BLEG776208
Marketsource
Supplier:   BIOLEGEND INC
Rat recombinant CNTF
Catalog # BLEG776208
Supplier:  BIOLEGEND INC
Marketsource
CAS Number:  
Rat recombinant CNTF

500 µG

CAS Number:  
Price
2 350,00 €
Log in for availability
1 * 500 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    500 µG
  • Cat. No.:
    BLEG776208
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    CNTF
  • Storage conditions:
    –20...–70 °C
  • Carrier-free:
    Y
  • Formulation:
    0,22 µm filtered protein solution is in 20 mM Tris, 150 mM NaCl, 1 mM EDTA, 2 mM CHAPS, 2 mM TCEP, pH 7,5
  • Protein synonyms:
    Ciliary neurotrophic factor
  • Concentration:
    0,2 mg/ml
  • Tested applications:
    BA
  • Species:
    Rat
  • Source:
    E. coli

Specifications