Human recombinant CEACAM-8/CD66b
100 µG
Catalog # BLEG778106
Marketsource
Supplier: BIOLEGEND INC
Human recombinant CEACAM-8/CD66b
Catalog # BLEG778106
Supplier: BIOLEGEND INC
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:100 µG
- Cat. No.:BLEG778106
- Protein/peptide type:Recombinant
- Protein/peptide name:CEACAM-8/CD66b
- Storage conditions:–20...–70 °C
- Carrier-free:Y
- Formulation:0,22 µm filtered protein solution is in PBS, pH 7,2
- Protein synonyms:CEACAM-8|Carcinoembryonic antigen CGM6|CD67 antigen|CEACAM8|CD66b|Carcinoembryonic antigen-related cell adhesion molecule 8|Nonspecific cross-reacting antigen NCA-95
- Concentration:0,2 mg/ml
- Tested applications:BA
- Species:Human
- Source:293E cells



