ContactUsLinkComponent
Recombinat Protein,Human IgG1 Fc Protein, His Tag,Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Glu 99 - Lys 330 (Accession P01857).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Catalog # ACRBIG1-H5225-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human IgG1 Fc Protein, His Tag,Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Glu 99 - Lys 330 (Accession P01857).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Catalog # ACRBIG1-H5225-1MG
Supplier:  ACROBIOSYSTEMS