Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HF520-EX-WBA3D1XX, LxBxH =160x100x61 mm, mit Display 2x Ausgang 4-20mA, 0-100%rf, 0-100%, 10-28V DC 1 * 1 items
Catalog # ROTRHF520-EX-WBA3D
Supplier:
ROTRONIC INSTRUMENTS
Expand image
HF520-EX-WBA3D1XX, LxBxH =160x100x61 mm, mit Display 2x Ausgang 4-20mA, 0-100%rf, 0-100%, 10-28V DC 1 * 1 items
Catalog # ROTRHF520-EX-WBA3D
Supplier:
ROTRONIC INSTRUMENTS