je 1000 mg/l in Salpetersaure 1 mol/l 1 * 500 mL
Catalog # KRAF27389.2000
Supplier: Bernd Kraft
Frequently Bought Together
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Maintenance solutions for electrodes
Supplier: Mettler - Toledo
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Hydrochloric acid 4 mol/l (4 N), VOLUSOL® volumetric solution
Supplier: VWR Chemicals
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Reagents for Karl Fischer titrations, Aquastar®, Supelco®
Supplier: Merck
Water standards for Karl Fischer titration, Hydranal™
Supplier: Honeywell Chemicals
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier: Merck