ContactUsLinkComponent
ICP-OES Wavelength Calibration Solution Concentrate 15 components Al 50ug/ml As 50ug/ml Ba 50ug/ml Cd 50ug/ml Co 50ug/ml Cr 50ug/ml Cu 50ug/ml Mn 50ug/ml Mo 50ug/ml Ni 50ug/ml Pb 50ug/ml Se 50ug/ml Sr 50ug/ml Zn 50ug/ml 5%. 1 * 100 mL
Artikelnr. CMPL6610030000.L1
ICP-OES Wavelength Calibration Solution Concentrate 15 components Al 50ug/ml As 50ug/ml Ba 50ug/ml Cd 50ug/ml Co 50ug/ml Cr 50ug/ml Cu 50ug/ml Mn 50ug/ml Mo 50ug/ml Ni 50ug/ml Pb 50ug/ml Se 50ug/ml Sr 50ug/ml Zn 50ug/ml 5%. 1 * 100 mL
Artikelnr. CMPL6610030000.L1
Frequently Bought Together