Order Entry
Germany
Orders LinkContactUsLinkComponent
Labels for BradyJet J2000 Lab printer

Labels for BradyJet J2000 Lab printer

Labels with 11 mm dot, B-7425J, J20-251-7425J, PP, White, J2000-Lab, 25×12 mm, 1500 labels/roll
Catalog # BRDY150033
Supplier:   Brady
Labels for BradyJet J2000 Lab printer
Catalog # BRDY150033
Supplier:  Brady
Labels for BradyJet J2000 Lab printer
Labels for BradyJet J2000 Lab printer

Labels with 11 mm dot, B-7425J, J20-251-7425J, PP, White, J2000-Lab, 25×12 mm, 1500 labels/roll

Price
178,47 €
Log in for availability
1 * 1 Rolle
Product Details & Documents

These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.

  • Designed to stay attached and remain legible
  • Resist liquid nitrogen, freezers, hot water baths and autoclaves

They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.

Specifications
  • Printer model:
    J2000-Lab
  • Type:
    B-7425J
  • Colour:
    White
  • Description:
    Labels with 11 mm dot
  • Dimensions:
    25×12 mm
  • Material:
    PP
  • Seq:
    05
  • Model:
    J20-251-7425J
  • Packed:
    1500 labels/roll
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.