Labels for BradyJet J2000 Lab printer
Printer labels, B-7425J, J20-256-7425J, PP, White, J2000-Lab, 33×15 mm, 1500 labels/roll
Artikelnummer BRDY150016
Hersteller: Brady
Wechseln zu:
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Dokumente
Brady Jet J2000 Laboratory Colour Label Printer
Produktliteratur
TDS B-7425J Inkjet Printable Propylene Label Stock TDS B-7425J Labels and Signs
Technisches Datenblatt
Spezifikationen:
- Printer model:J2000-Lab
- Type:B-7425J
- Colour:White
- Description:Printer labels
- Dimensions:33×15 mm
- Material:PP
- Seq:09
- Model:J20-256-7425J
- Packed:1500 labels/roll



