Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
5-way header, M16×1, The 5-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Artikelnummer HUBE341892
Hersteller:   HUBER
Accessories for circulators
Artikelnummer HUBE341892
Hersteller:  HUBER
Accessories for circulators
5-way header, M16×1, The 5-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Preis
510,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    5-way header, M16×1
  • Seq:
    16
  • For:
    The 5-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.