Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
4-way header, M16×1, The 4-way header with an universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Catalog # HUBE341871
Supplier:   HUBER
Accessories for circulators
Catalog # HUBE341871
Supplier:  HUBER
Accessories for circulators
4-way header, M16×1, The 4-way header with an universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
Note: The manifold must not be mounted on the unit.
Price
390,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    4-way header, M16×1
  • Seq:
    03
  • For:
    The 4-way header with an universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit.
    Note: The manifold must not be mounted on the unit.