Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Q. Station 101 1 Pieces 2,420.00 2,420.00 High-performance controller for highly data rates: 8 Measuring channels (2 channels per RS -485) 100 kh
Catalog # MSPP290124
Supplier:
MARKET SOURCE PART PROCESS
Expand image
Q. Station 101 1 Pieces 2,420.00 2,420.00 High-performance controller for highly data rates: 8 Measuring channels (2 channels per RS -485) 100 kh
Catalog # MSPP290124
Supplier:
MARKET SOURCE PART PROCESS