Orders LinkContactUsLinkComponent
Gel documentation system, Quantum CX5 Edge
Gel documentation system, Quantum CX5 Edge
Gel documentation system, Quantum CX5 Edge, with Epi UV for TLC
Catalog # VILB101196701
Supplier:   Vilber Lourmat
Gel documentation system, Quantum CX5 Edge
Catalog # VILB101196701
Supplier:  Vilber Lourmat
Gel documentation system, Quantum CX5 Edge
Gel documentation system, Quantum CX5 Edge
Gel documentation system, Quantum CX5 Edge, with Epi UV for TLC
Price
10 550,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The Quantum CX5 Edge gel documentation system is an ultra-modern, fully automated, yet compact system with innovative features and superior technology. With just one click, it allows the recording of images of the highest quality ('One-Button-Imaging').
  • Easy to operate, standalone
  • 'Scientific-grade' camera with 5 MP resolution and 6× automated zoom lens
  • Robust and light-proof all-metal darkroom
  • Including extendable transilluminator (optional as UV, SUPER-BRIGHT, blue and white light) usable with only one hand and interchangeable
  • Unprecedented sensitivity on a receiving surface of up to 21×26 cm
For maximum flexibility in fluorescence applications an optional Epi-UV illumination unit can also be installed. Thus the system can be perfectly adapted to experimental conditions.
Ordering information: The powerful BIO-VISION software (included) offers extensive possibilities of recording, display, analysis and output, and is clearly designed and easy to use.
Specifications
  • Description:
    Gel documentation system, Quantum CX5 Edge, with Epi UV for TLC
  • Seq:
    01
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Power cord with EU plug
    Power cord with EU plug
    733-2833
    Log in for availability
    1 * 1 ST - 6,20 €