ContactUsLinkComponent
Conventional stain used for Hematoxylin and Eosin (H&E) staining in histology and cytology procedures. Ready to use formulation opt imized for histology and cytology procedures where reproducible results are important. null
Catalog # GENO786-1059
Supplier:  G-Biosciences
Conventional stain used for Hematoxylin and Eosin (H&E) staining in histology and cytology procedures. Ready to use formulation opt imized for histology and cytology procedures where reproducible results are important. null
Catalog # GENO786-1059
Supplier:  G-Biosciences