ContactUsLinkComponent
Aqueous Eosin Y Solution [0.5%], a conventional stain used for Hematoxylin and Eosin (H&E) staining in histology and cytology proced ures. Ready to use formulation optimized for histology and cytology procedures where reproducible results are importa 1 * 1 L
Catalog # GENO786-1070
Supplier:  G-Biosciences
Aqueous Eosin Y Solution [0.5%], a conventional stain used for Hematoxylin and Eosin (H&E) staining in histology and cytology proced ures. Ready to use formulation optimized for histology and cytology procedures where reproducible results are importa 1 * 1 L
Catalog # GENO786-1070
Supplier:  G-Biosciences