[EN]SODIUM HYDROXIDE 0,4 MOL/L IN METHAN 1 * 2,5 L
Catalog # PRLS89955.320
Supplier: PROLABO CMP
Frequently Bought Together
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
VWR® Q Path® DC3, Decalcifier Rapid-Acting
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Standards, calibration for LCK cuvettes, Addista
Supplier: Hach
Soda lime, AnalaR NORMAPUR® analytical reagent, with indicator
Supplier: VWR Chemicals
Reagents for nitrate determination using Lovibond® MD photometers
Supplier: Lovibond
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Orthoboric acid 40 g/l in aqueous solution for Kjeldahl determinations, with indicator
Supplier: VWR Chemicals
Reagents for iron determination using Lovibond® MD Photometers
Supplier: Lovibond
Test kits, photometric, nickel, LCK cuvette tests
Supplier: Hach
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for determination of silica
Supplier: Hach
Powder pillows, chlorine, free and total
Supplier: Hach
Bromocresol green-methyl red solution mixed indicator, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY