Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HELIFLEX+WAX 30M X 0.32MM X 0.25UM 1 * 1 items
Catalog # HISM8635275
Supplier:
HICHROM
Expand image
HELIFLEX+WAX 30M X 0.32MM X 0.25UM 1 * 1 items
Catalog # HISM8635275
Supplier:
HICHROM