ContactUsLinkComponent
Bottletop dispenser Dispensette® S Organic, Analog-adjustable, DE-M marking, 0,5-5 ml, with recirculation valve dakks 1 * 1 items
Catalog # BRNDDAKKS4630131
Supplier:  Brand
Bottletop dispenser Dispensette® S Organic, Analog-adjustable, DE-M marking, 0,5-5 ml, with recirculation valve dakks 1 * 1 items
Catalog # BRNDDAKKS4630131
Supplier:  Brand