ContactUsLinkComponent
Hydrothermal Synthesis Reactor, Capacity:500ml, Safe Temperature:180℃, Working pressure:Less than3 Mpa Shell Material:SS304 (stainle ss steel), Liner material,color:Teflon/white, Temperature heating and cooling speed:Less than 5℃/ mi
  IBSAHS-REACTOR
Hydrothermal Synthesis Reactor, Capacity:500ml, Safe Temperature:180℃, Working pressure:Less than3 Mpa Shell Material:SS304 (stainle ss steel), Liner material,color:Teflon/white, Temperature heating and cooling speed:Less than 5℃/ mi
  IBSAHS-REACTOR