Order Entry
Germany
Orders LinkContactUsLinkComponent
 

UV transilluminators, multibrand

TCP-20.MC V1, 5×8 / 6×8 W, 200×200 mm, 312 / 254 nm
  VILB216120171
UV transilluminators, multibrand
  VILB216120171
UV transilluminators, multibrand
UV transilluminators, multibrand

TCP-20.MC V1, 5×8 / 6×8 W, 200×200 mm, 312 / 254 nm

 
 
   
 

This UV transilluminator accommodates a wide range of applications requiring different wavelengths for both visualisation and documentation.

  • UV protection shield
  • Two UV wavelengths in one single transilluminator
  • Super-Bright filter to stop the visible light from UV tubes for an unbeatable viewing field

15-Watt W×D×H: 470×330×150 mm

 
  • Model:
    TCP-20.MC V1
  • Wavelength:
    312 / 254 nm
  • Filter size:
    200×200 mm
  • Output:
    5×8 / 6×8 W