Orders LinkContactUsLinkComponent
UV transilluminators
UV transilluminators
Super-Bright UV-Pad Edge 26LMX, 210×260 mm, 312/365 nm
Catalog # VILB280126141
Supplier:   Vilber Lourmat
UV transilluminators
Catalog # VILB280126141
Supplier:  Vilber Lourmat
UV transilluminators
UV transilluminators
Super-Bright UV-Pad Edge 26LMX, 210×260 mm, 312/365 nm
Price
2 350,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Model:
    Super-Bright UV-Pad Edge 26LMX
  • Wavelength:
    312/365 nm
  • Seq:
    04
  • Filter size:
    210×260 mm