Orders LinkContactUsLinkComponent
UV transilluminators
UV transilluminators
UV-Pad Edge 26LM, 210×260 mm, 312/365 nm
Catalog # VILB280126161
Supplier:   Vilber Lourmat
UV transilluminators
Catalog # VILB280126161
Supplier:  Vilber Lourmat
UV transilluminators
UV transilluminators
UV-Pad Edge 26LM, 210×260 mm, 312/365 nm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Model:
    UV-Pad Edge 26LM
  • Wavelength:
    312/365 nm
  • Seq:
    05
  • Filter size:
    210×260 mm