Order Entry
Germany
Orders LinkContactUsLinkComponent
 

UV transilluminators

Super-Bright UV-Pad Edge 26MX, 210×260 mm, 312 nm
Catalog # 733-2834
Supplier:   Vilber Lourmat
UV transilluminators
Catalog # 733-2834
Supplier:  Vilber Lourmat
UV transilluminators
UV transilluminators

Super-Bright UV-Pad Edge 26MX, 210×260 mm, 312 nm

Price
1 810,00 €
Log in for availability
Product Details & Documents
Specifications
  • Model:
    Super-Bright UV-Pad Edge 26MX
  • Wavelength:
    312 nm
  • Seq:
    01
  • Filter size:
    210×260 mm