Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
OV-17 GC COLUMN 30M x 0.53MM x 1.00UM 1 * 1 items
Catalog # HISM1730-5304
Supplier:
HICHROM
Expand image
OV-17 GC COLUMN 30M x 0.53MM x 1.00UM 1 * 1 items
Catalog # HISM1730-5304
Supplier:
HICHROM