Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution 25 - 27%, NORMATOM® for trace analysis

Ammonia solution 25 - 27%, NORMATOM® for trace analysis

0,5 L
Catalog # 85693.270
Corrosive
Irritant
Supplier:   VWR Chemicals
Ammonia solution 25 - 27%, NORMATOM® for trace analysis
Catalog # 85693.270
Supplier:  VWR Chemicals
CAS Number:  1336-21-6
Ammonia solution 25 - 27%, NORMATOM® for trace analysis
Ammonia solution 25 - 27%, NORMATOM® for trace analysis

0,5 L

CAS Number:  1336-21-6
Price
111,00 €
Log in for availability
1 * 0,5 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • UN:2672
  • ADR:8,III
Specifications

Specification Test Results

  • Assay (NH3):
    25 - 27% w/w
  • Colouration:
    max. 10 APHA
  • Cl (Chloride):
    max. 0.5 ppm
  • PO4 (Phosphate):
    max. 0.01 ppm
  • SO4 (Sulphate):
    max. 1 ppm
  • Ag (Silver):
    max. 0.5 ppb
  • Al (Aluminum):
    max. 1 ppb
  • As (Arsenic):
    max. 1 ppb
  • Au (Gold):
    max. 0.5 ppb
  • Ba (Barium):
    max. 0.1 ppb
  • Be (Beryllium):
    max. 0.1 ppb
  • Bi (Bismuth):
    max. 0.1 ppb
  • Ca (Calcium):
    max. 1 ppb
  • Cd (Cadmium):
    max. 0.5 ppb
  • Ce (Cerium):
    max. 0.1 ppb
  • Co (Cobalt):
    max. 0.5 ppb
  • Cr (Chromium):
    max. 0.5 ppb
  • Cs (Cesium):
    max. 0.1 ppb
  • Cu (Copper):
    max. 0.5 ppb
  • Dy (Dysprosium):
    max. 0.1 ppb
  • Er (Erbium):
    max. 0.1 ppb
  • Eu (Europium):
    max. 0.1 ppb
  • Fe (Iron):
    max. 1 ppb
  • Ga (Gallium):
    max. 0.1 ppb
  • Gd (Gadolinium):
    max. 0.1 ppb
  • Ge (Germanium):
    max. 0.1 ppb
  • Hf (Hafnium):
    Information Only
  • Hg (Mercury):
    max. 0.2 ppb
  • Ho (Holmium):
    max. 0.1 ppb
  • In (Indium):
    max. 0.1 ppb
  • K (Potassium):
    max. 1 ppb
  • La (Lanthanum):
    max. 0.1 ppb
  • Li (Lithium):
    max. 0.1 ppb
  • Lu (Lutetium):
    max. 0.1 ppb
  • Mg (Magnesium):
    max. 1 ppb
  • Mn (Manganese):
    max. 0.5 ppb
  • Mo (Molybdenum):
    max. 0.5 ppb
  • Na (Sodium):
    max. 1 ppb
  • Nb (Niobium):
    max. 0.1 ppb
  • Nd (Neodymium):
    max. 0.1 ppb
  • Ni (Nickel):
    max. 0.5 ppb
  • Pb (Lead):
    max. 0.1 ppb
  • Pd (Palladium):
    Information Only
  • Pr (Praseodymium):
    max. 0.1 ppb
  • Pt (Platinum):
    Information Only
  • Rb (Rubidium):
    max. 0.1 ppb
  • Re (Rhenium):
    Information Only
  • Rh (Rhodium):
    max. 0.5 ppb
  • Ru (Ruthenium):
    Information Only
  • Sb (Antimony):
    max. 0.5 ppb
  • Sc (Scandium):
    max. 0.1 ppb
  • Se (Selenium):
    max. 1 ppb
  • Sm (Samarium):
    max. 0.1 ppb
  • Sn (Tin):
    max. 0.5 ppb
  • Sr (Strontium):
    max. 0.1 ppb
  • Tb (Terbium):
    max. 0.1 ppb
  • Te (Tellurium):
    max. 0.1 ppb
  • Th (Thorium):
    max. 0.1 ppb
  • Ti (Titanium):
    max. 0.5 ppb
  • Tl (Thallium):
    max. 0.1 ppb
  • Tm (Thulium):
    max. 0.1 ppb
  • U (Uranium):
    max. 0.1 ppb
  • V (Vanadium):
    max. 0.5 ppb
  • W (Tungsten):
    max. 0.1 ppb
  • Y (Yttrium):
    max. 0.1 ppb
  • Yb (Ytterbium):
    max. 0.1 ppb
  • Zn (Zinc):
    max. 0.5 ppb
  • Zr (Zirconium):
    max. 0.1 ppb

Specifications

  • Pk:
    0,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.