Order Entry
Germany
Orders LinkContactUsLinkComponent
Creatinine NF

Creatinine NF

100 g
Catalog # SPCMC1403-100GM
Supplier:   Spectrum Chemical
Creatinine NF
Catalog # SPCMC1403-100GM
Supplier:  Spectrum Chemical
CAS Number:  60-27-5
Creatinine NF

100 g

CAS Number:  60-27-5
Price
290,00 €
Log in for availability
1 * 100 g
Product Details & Documents

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-466-7
Specifications

Specification Test Results

  • Assay:
    98.5% Min.

Specifications

  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.