Order Entry
Germany
Orders LinkContactUsLinkComponent
UV transilluminators

UV transilluminators

Skylight-Pad Edge 20 WL, 200×200 mm, 470 nm
Artikelnummer VILB280120181
Hersteller:   Vilber Lourmat
UV transilluminators
Artikelnummer VILB280120181
Hersteller:  Vilber Lourmat
UV transilluminators
UV transilluminators

Skylight-Pad Edge 20 WL, 200×200 mm, 470 nm

Preis
1 850,00 €
Log in for availability
Product Details & Documents
Specifications
  • Model:
    Skylight-Pad Edge 20 WL
  • Wavelength:
    470 nm
  • Seq:
    06
  • Filter size:
    200×200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.