Skip to Footer
Erweiterte Suche
Erweiterte Suche
Sicherheitsdatenblatt (SDS)
Zertifikate
Herstellerdaten
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Kit Poly(styrene) high, 12 x 1.0 g, Mp 682 - 2 520 000 Da 1 * 1 KIT
Catalog # POSSPSKITH-11
Supplier:
POLYMER STANDARDS SERVICE
Expand image
Kit Poly(styrene) high, 12 x 1.0 g, Mp 682 - 2 520 000 Da 1 * 1 KIT
Catalog # POSSPSKITH-11
Supplier:
POLYMER STANDARDS SERVICE