Order Entry
Germany
Orders LinkContactUsLinkComponent
D(-)-Lactic acid

D(-)-Lactic acid

Bottle, 50 g
Catalog # APOSOR960721-50G
Irritant
Supplier:   Apollo Scientific
D(-)-Lactic acid
Catalog # APOSOR960721-50G
Supplier:  Apollo Scientific
CAS Number:  10326-41-7
D(-)-Lactic acid

Bottle, 50 g

CAS Number:  10326-41-7
Price
170,00 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
  • CAS Number:10326-41-7
  • Formula:C₃H₆O₃
  • MDL Number:MFCD00068311
  • Molecular Weight:90.08 g/mol
  • Storage Temperature:Freezer

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 g

Specifications