Order Entry
Germany
Orders LinkContactUsLinkComponent
 

(R)-(-)-2-Butanol 99+%

Bottle, 1 g
Catalog # APOSOR304038-1G
Flammable
Irritant
Supplier:   Apollo Scientific
(R)-(-)-2-Butanol 99+%
Catalog # APOSOR304038-1G
Supplier:  Apollo Scientific
CAS Number:  14898-79-4
(R)-(-)-2-Butanol 99+%

Bottle, 1 g

CAS Number:  14898-79-4
Price
79,60 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol
  • CAS Number:14898-79-4
  • Formula:C₄H₁₀O
  • MDL Number:MFCD00064280
  • Molecular Weight:74.12 g/mol
  • Boiling Pt:99…100 °C
  • Melting Pt:–114 °C
  • Density:0.806 g/cm³ (25 °C)
  • Flash Pt:26 °C
  • Storage Temperature:Ambient
  • UN:1120
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications