Methyl 2-methyl-6-nitrobenzoate
Bottle, 1 g
Catalog # APOSOR301028-1G
Supplier: Apollo Scientific
Methyl 2-methyl-6-nitrobenzoate
Catalog # APOSOR301028-1G
Supplier: Apollo Scientific
CAS Number: 61940-22-5
Jump to:
Product Details & Documents
Synonyms: Methyl 6-nitro-o-toluate, 2-(Methoxycarbonyl)-3-nitrotoluene, 2-(Methoxycarbonyl)-3-methylnitrobenzene
- CAS Number:61940-22-5
- Formula:C₉H₉NO₄
- MDL Number:MFCD11042453
- Molecular Weight:195.172 g/mol
- Melting Pt:50 °C
- Storage Temperature:Raumtemperatur
Specifications
Specifications
- Packung:Bottle
- VE:1 g



