Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Dropping pipettes, disposable

Disposable pipette, 2,4 ml, 155 mm
Catalog # BURK7555-0003
Supplier:   BURKLE
Dropping pipettes, disposable
Catalog # BURK7555-0003
Supplier:  BURKLE
Dropping pipettes, disposable
Dropping pipettes, disposable

Disposable pipette, 2,4 ml, 155 mm

Price
25,90 €
Log in for availability
1 * 100 ST
Product Details & Documents

Made of LDPE.

  • Ungraduated
Specifications
  • Capacity:
    2,4 ml
  • Description:
    Disposable pipette
  • Length:
    155 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.