ContactUsLinkComponent
Transmitter M400 4-Wire, Pharmaceutical The multi-parameter M400 transmitter series features Intelligent Sensor Management (ISM®) technology for pH / ORP, dissolved oxygen, dissolved carbon dioxide and conductivity measurement. 1 * 1 items
Catalog # METN30374112
Supplier:  Mettler - Toledo
Transmitter M400 4-Wire, Pharmaceutical The multi-parameter M400 transmitter series features Intelligent Sensor Management (ISM®) technology for pH / ORP, dissolved oxygen, dissolved carbon dioxide and conductivity measurement. 1 * 1 items
Catalog # METN30374112
Supplier:  Mettler - Toledo
Frequently Bought Together