Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
William’s Medium E, w/o: L-Glutamine, w/o: Phenol red, w: 2.24 g/L NaHCO3, 500 ml 1 * 500 mL
Catalog # PANBP04-29510
Supplier:
PAN BIOTECH
Expand image
William’s Medium E, w/o: L-Glutamine, w/o: Phenol red, w: 2.24 g/L NaHCO3, 500 ml 1 * 500 mL
Catalog # PANBP04-29510
Supplier:
PAN BIOTECH