Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DMEM w: 1,0 g/l Glucose, w: stab. Glutamine, w: Sodium pyruvate, w: 3,7 g/l NaHCO3 1 * 500 mL
Catalog # PANBP04-02500
Supplier:
PAN BIOTECH
Expand image
DMEM w: 1,0 g/l Glucose, w: stab. Glutamine, w: Sodium pyruvate, w: 3,7 g/l NaHCO3 1 * 500 mL
Catalog # PANBP04-02500
Supplier:
PAN BIOTECH