Orders LinkContactUsLinkComponent
Accessories
Accessories
Lysing tube, skirted, PE, with red cap, animal tissue
Catalog # 431-0267
Supplier:   OHAUS
Accessories
Catalog # 431-0267
Supplier:  OHAUS
Accessories
Accessories
Lysing tube, skirted, PE, with red cap, animal tissue
Price
402,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Specifications
  • Description:
    Lysing tube, skirted, PE, with red cap
  • Seq:
    06
  • For:
    animal tissue