VWR® Basic, Bouffant Caps, Latex-Free
Green, 610 mm
Catalog # 113-8885
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Free from natural rubber latex
- Available in a wide range of colours
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.
Documents
CoC VWR® Basic Bouffant Caps.pdf
Conformity declaration
CoC_VWRI113-1459_03200637 COC_VWRI113-1459_03221002 COC_VWRI113-1459_03221003 COC_VWRI113-1459_03231100 COC_VWRI113-1459_03231364
More
Food Contact Certificate
Additional Product Details
VWR® Non Regulated Disposables for Production
Product Literature
Specifications
- Ø:610 mm
- Colour:Green
- Seq:08



