Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR® Basic, Overalls, with Hood

VWR® Basic, Overalls, with Hood

Blue, 3XL
Catalog # 113-9025
Supplier:   VWR Collection
VWR® Basic, Overalls, with Hood
Catalog # 113-9025
Supplier:  VWR Collection
VWR® Basic, Overalls, with Hood
VWR® Basic, Overalls, with Hood

Blue, 3XL

Price
289,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

These disposable overalls, with hood, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.

  • Hood
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex

Weight: 40 g/m²

Packaging: 1 piece per bag, 50 bags per case.

Specifications
  • Clothing Size:
    3XL
  • Colour:
    Blue
  • Seq:
    12
  • Cat. No.:
    113-9025
  • Garment application:
    General Purpose|Laboratory
  • Weight:
    40 g/m²