VWR® Basic, Overalls, with Hood
Blue, S
Catalog # 113-9020
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable overalls, with hood, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Hood
- Zip closure (one direction)
- Elasticated wrists
- Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Documents
Specifications
- Clothing Size:S
- Colour:Blue
- Seq:07
- Cat. No.:113-9020
- Garment application:General Purpose|Laboratory
- Weight:40 g/m²