Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR® Basic, Overalls, with Hood

VWR® Basic, Overalls, with Hood

White, 3XL
Catalog # 113-9019
Supplier:   VWR Collection
VWR® Basic, Overalls, with Hood
Catalog # 113-9019
Supplier:  VWR Collection
VWR® Basic, Overalls, with Hood
VWR® Basic, Overalls, with Hood

White, 3XL

Price
289,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

These disposable overalls, with hood, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.

  • Hood
  • Zip closure (one direction)
  • Elasticated wrists
  • Free from natural rubber latex

Weight: 40 g/m²

Packaging: 1 piece per bag, 50 bags per case.

Specifications
  • Clothing Size:
    3XL
  • Colour:
    White
  • Seq:
    06
  • Cat. No.:
    113-9019
  • Garment application:
    General Purpose|Laboratory
  • Weight:
    40 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.