Orders LinkContactUsLinkComponent
VWR® Basic, Overalls, Dual-Zip Closure
VWR® Basic, Overalls, Dual-Zip Closure
White, M
Catalog # 113-9010
Supplier:   VWR Collection
VWR® Basic, Overalls, Dual-Zip Closure
Catalog # 113-9010
Supplier:  VWR Collection
VWR® Basic, Overalls, Dual-Zip Closure
VWR® Basic, Overalls, Dual-Zip Closure
White, M
Price
132,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
These disposable overalls, with collar, are made of non woven, spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • Collar
  • Zip closure (dual-direction)
  • Elasticated wrists
  • Free from natural rubber latex
Weight: 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Specifications
  • Clothing Size:
    M
  • Colour:
    White
  • Seq:
    01
  • Cat. No.:
    113-9010
  • Garment application:
    General Purpose|Laboratory|Industrial
  • Closure type:
    Zip closure
  • Weight:
    40 g/m²