VWR® Basic Visitor Coats, Velcro® Closure
Blue, L, 104 cm
113-8966
These disposable coats, with collar, are made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- VELCRO® closure
- Elastic wrists
- Free from natural rubber latex
- Available in white and blue
Weight: 30 g/m²
: 1 piece per bag, 50 bags per case.
- Clothing Size:L
- Colour:Blue
- Length:104 cm
- Gender:Unisex
- Closure type:Hook-and-loop / VELCRO®
- Garment application:Laboratory
- Weight:30 g/m²
- Cat. No.:113-8966
- Pockets:Without pockets



