Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR® Basic Visitor Coats, Velcro® Closure

VWR® Basic Visitor Coats, Velcro® Closure

White, XXL, 112 cm
Catalog # 113-8960
Supplier:   VWR Collection
VWR® Basic Visitor Coats, Velcro® Closure
Catalog # 113-8960
Supplier:  VWR Collection
VWR® Basic Visitor Coats, Velcro® Closure
VWR® Basic Visitor Coats, Velcro® Closure

White, XXL, 112 cm

Price
181,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

These disposable coats, with collar, are made from non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.

  • VELCRO® closure
  • Elastic wrists
  • Free from natural rubber latex
  • Available in white and blue

Weight: 30 g/m²

Packaging: 1 piece per bag, 50 bags per case.

Specifications
  • Clothing Size:
    XXL
  • Colour:
    White
  • Length:
    112 cm
  • Cat. No.:
    113-8960
  • Garment application:
    Laboratory
  • Gender:
    Unisex
  • Pockets:
    Without pockets
  • Closure type:
    Hook-and-loop / VELCRO®
  • Weight:
    30 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.